missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DNASE1L1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13653-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes a deoxyribonuclease protein that shows high sequence similarity to DNase I. The encoded protein is localized to the endoplasmic reticulum and modified by N-linked glycosylation. Alternate transcriptional splice variants encoding the same protein have been observed.Specifications
| DNASE1L1 | |
| Polyclonal | |
| Unconjugated | |
| DNASE1L1 | |
| Deoxyribonuclease 1 like 1, deoxyribonuclease I like 1, DNAS1L1, DNase I like 1, DNase X, DNASE1L1, DNASEX, DNL1L, Muscle specific DNase I like, XIB | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 1774, 363522, 69537 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.21 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P49184, Q2QDE7, Q9D7J6 | |
| DNASE1L1 | |
| DNASE1L1 Fusion Protein Ag4222 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction