missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ DNASE1 (Bovine) Recombinant Protein

Product Code. 16181830
Click to view available options
Quantity:
10,000 U
Unit Size:
10000 units
This item is not returnable. View return policy

Product Code. 16181830

Brand: Abnova™ P5311.10000Units

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used for Enzyme

#REF!

Specifications

Accession Number P00639
For Use With (Application) Enzyme Activity
Gene ID (Entrez) 282217
Molecular Weight (g/mol) 31.3kDa
Name DNASE1 (Bovine) Recombinant Protein
Preparation Method Yeast expression system
Quantity 10,000 U
Storage Requirements Store at 4°C on dry atmosphere.
Regulatory Status RUO
Common Name DNASE1
Gene Symbol DNASE1
Assays Deoxyribonuclease I Assay; Removing genomic DNA from RNA preparations prior to RT-PCR; Degradation of DNA templates after transcription reactions; Removing unwanted DNA from samples prior to Northern blotting; Removing DNA during biopharma and bioprocessing procedures.
Biological Activity ≥5000 units per mg protein. One Unit causes an increase in absorbance at 260nm of 0.001 per minute per ml at 25°C when acting upon highly polymerized DNA at pH 5.0.
Species Yeast
Recombinant Recombinant
Protein Tag None
Expression System Yeast expression system
Form Lyophilized
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.