missing translation for 'onlineSavingsMsg'
Learn More

DNase-, RNase-Free Water, MP Biomedicals™

Product Code. 11430615 Shop All MP Biomedicals Products
Change view
Click to view available options
:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
11430615 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11430615 Supplier MP Biomedicals Supplier No. 04821932

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Suitable for the investigation of nucleic acid and protein synthesis.

  • Grade: DNase- RNase-free
  • Key applications: Investigation of nucleic acid and protein synthesis
  • Clear, colorless, liquid
  • 0.2 μ filtered (sterile)
TRUSTED_SUSTAINABILITY

Specifications

CAS 7732-18-5
Color Clear, Colorless
Boiling Point 100°C
DNase DNase-free
Molecular Formula H2O
MDL Number MFCD00011332
UN Number 0000
Merck Index 13.10098
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
SMILES O
IUPAC Name water
Molecular Weight (g/mol) 18.02
Grade Molecular Biology
Density 1 g/cm3 (4°C)
Quantity 500 mL
Refractive Index n20/D 1.34(Lit.)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.