missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DNase II Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5115136
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total DNase II.
DNase II is a member of the DNase family. The protein, located in the lysosome, hydrolyzes DNA under acidic conditions and mediates the breakdown of DNA during erythropoiesis and apoptosis. Two codominant alleles have been characterized, DNASE2*L (low activity) and DNASE2*H (high activity), that differ at one nucleotide in the promoter region. The DNASE2*H allele is represented in this record.
Specifications
| DNase II | |
| Polyclonal | |
| Unconjugated | |
| DNASE2 | |
| acid DNase; deoxyribonuclease; Deoxyribonuclease II alpha; deoxyribonuclease II, lysosomal; deoxyribonuclease-2-alpha; DNase II alpha; DNase II, lysosomal; DNASE2; Dnase2a; DNL; Dnl2; lysosomal DNase II; R31240_2 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 171575, 1777 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide | |
| O00115, Q9QZK8 | |
| DNASE2 | |
| A synthesized peptide derived from human DNASE2(Accession O00115), corresponding to amino acid residues E209-T259. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction