missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DNASE1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA552731
This item is not returnable.
View return policy
Description
Immunogen sequence: YVRPSQWSSI RLWTSPTFQW LIPDSADTTA TPTHCAYDRI VVAGMLLRGA VVPDSALPFN FQAAYGLSDQ LAQAISDHY Highest antigen sequence identity to the following orthologs: Mouse - 81%, Rat - 78%.
Specifications
| DNASE1 | |
| Polyclonal | |
| Unconjugated | |
| DNASE1 | |
| deoxyribonuclease 1; deoxyribonuclease I; Deoxyribonuclease-1; DNase I; DNase I, lysosomal; DNASE1; DNL1; Dornase alfa; DRNI; epithelial lens deoxyribonuclease I; human urine deoxyribonuclease I; pancreatic deoxyribonuclease I | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 1773 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.20 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P24855 | |
| DNASE1 | |
| Recombinant protein corresponding to Human DNase I. Recombinant protein control fragment (Product #RP-90198). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction