missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies DNase I, ACF

Product Code. 17177501
Encompass_Preferred
Change view
Click to view available options
Quantity:
100,000 U
25,000 U
500,000 U
Unit Size:
1 set
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17177501 500,000 U 1 set
17149281 25,000 U 1 set
17159281 100,000 U 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17177501 Supplier STEMCELL Technologies Supplier No. 1000273

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Animal component-free DNase I for digestion of DNA

  • Deoxyribonuclease I (DNase I), Animal Component-Free (ACF) is obtained from cultures free of animal-derived materials and purified by chromatography
  • DNase I is an endonuclease consisting of a single glycosylated polypeptide chain with two disulfide bonds
  • DNase is often included in tissue dissociation protocols to digest DNA that has leaked into the dissociation medium as a result of cell damage
  • DNase I, ACF preferentially cleaves phosphodiester linkages adjacent to pyrimidine nucleotides in both single- and double-stranded DNA, yielding polynucleotides with 5'-phosphate and 3'-hydroxyl groups (Bernardi et al.) DNase I has been used for the digestion of human tissues such as microglia (Klegeris & McGeer), cartilage (Dunham & Koch), colon (Fukushima & Fiocchi), epithelium l (Fukushima & Fiocchi), liver (Vatakis et al.), lung (Fujino et al.), neural (Fuja et al.), and stem cells (Kusuma et al.)

Specifications

For Use With (Application) Digestion of DNA
Quantity 500,000 U
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.