missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ DNase (Bovine)

Product Code. 16131750
Click to view available options
Quantity:
1 Tub
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 16131750

Brand: Abnova™ P5200.1Tub

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used for Enzyme

#REF!

Specifications

Accession Number P00639
For Use With (Application) Enzyme Activity
Gene ID (Entrez) 282217
Molecular Weight (g/mol) 31.3kDa
Name DNase (Bovine)
Preparation Method Native protein purified from Bovine Pancreas
Quantity 1 Tub
Storage Requirements Store at 4°C. Reconstituted with 0.5 ml of Earle's Balanced Salt Solution (EBSS) or equivalent yields a solution of 2000 units/ml (1 mg/ml).
Regulatory Status RUO
Common Name DNASE1
Gene Symbol DNASE1
Assays Deoxyribonuclease I Assay; DNA removal in primary cell isolation to decreases viscosity providing better yields; DNA removal in bioprocessing applications.
Biological Activity ≥1,000 units per vial. One Unit causes an increase in absorbance at 260 nm of 0.001 per minute per ml at 25°C when acting upon highly polymerized DNA at pH 5.0.
Species Bovine
Protein Tag None
Expression System Native protein purified from Bovine Pancreas
Form Lyophilized
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.