missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
DNALI1 Polyclonal antibody specifically detects DNALI1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antigen | DNALI1 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500 |
| Formulation | PBS (pH 7.2) and 40% Glycerol |
| Gene Alias | dJ423B22.5, dynein, axonemal, light intermediate chain 1, dynein, axonemal, light intermediate polypeptide 1, hp28axonemal dynein light intermediate polypeptide 1, Inner dynein arm light chain, axonemal, inner dynein arm, homolog of clamydomonas, P28dJ423B22.5 (axonemal dynein light chain (hp28)) |
| Host Species | Rabbit |
| Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids: CWVATLASAMIPPADSLLKYDTPVLVSRNTEKRSPKARLLKVSPQQPGPSGSAPQPPKTKLPSTPCVPDPTKQ |
| Purification Method | Immunogen affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?