missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ DNAL1 Polyclonal Antibody
GREENER_CHOICE

Product Code. 12828752
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12828752 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. Se returpolicy
Produktkod. 12828752 Leverantör Invitrogen™ Leverantörsnummer PA520573

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

Rabbit Polyclonal Antibody

A suggested positive control is NIH-3T3 cell lysate. PA5-20573 can be used with blocking peptide PEP-0693.

DNAL1 was identified as a potential candidate gene for primary ciliary dyskinesia (PCD), a genetically heterologous disorder characterized by chronic infections of the upper and lower airways that often leads to permanent lung damage, randomization of left/right body symmetry, and reduced fertility. DNAL1 is reported to be expressed solely in tissues carrying motile cilia for flagella and interacts with DNAH5, a protein that when mutated has been shown to result in PCD. It has been suggested that DNAL1 serves a regulatory function for DNAH5 activity in outer dynein arms of sperm flagella, respiratory cilia, and ependymal cilia. DNAL1 has also been recently identified as an HIV dependency factor (HDF), suggesting that DNAL1 may be an important drug target in HIV treatment. At least two isoforms of DNAL1 are known to exist.
TRUSTED_SUSTAINABILITY

Specifikationer

Antigen DNAL1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene Dnal1
Gene Accession No. Q05A62, Q4LDG9
Gene Alias 1700010H15Rik; AW121714; axonemal dynein light chain 1; C14orf168; CILD16; DNAL1; Dnalc1; dynein axonemal light chain 1; dynein light chain 1, axonemal; dynein, axonemal, light chain 1; E330027P08Rik; LC1
Gene Symbols Dnal1
Host Species Rabbit
Immunogen A 17 amino acid peptide from near the carboxy terminus of human DNAL1.
Purification Method Antigen Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 105000, 685664, 83544
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Visa mer Visa mindre
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.