missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DNAJC8 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£248.00 - £470.00
Specifications
| Antigen | DNAJC8 |
|---|---|
| Dilution | Western Blot 0.4 ug/ml, Simple Western 1:5, Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200 |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18451391
|
Novus Biologicals
NBP1-87015-25ul |
25 μL |
£248.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18286106
|
Novus Biologicals
NBP1-87015 |
0.1 mL |
£470.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
DNAJC8 Polyclonal specifically detects DNAJC8 in Human samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| DNAJC8 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| DnaJ (Hsp40) homolog, subfamily C, member 8, dnaJ homolog subfamily C member 8, HSPC331, SPF31Splicing protein spf31 | |
| DNAJC8 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Western Blot 0.4 ug/ml, Simple Western 1:5, Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 22826 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:ELEIKRKEREAKEMHERKRQREEEIEAQEKAKREREWQKNFEESRDGRVDSWRNFQANTKGKKEKKNRTFLRPPKVK | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title