missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DNAJC8 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-97847
This item is not returnable.
View return policy
Description
DNAJC8 Polyclonal specifically detects DNAJC8 in Human samples. It is validated for Western Blot, Immunoprecipitation.
Specifications
| DNAJC8 | |
| Polyclonal | |
| Unconjugated | |
| TBS and 0.1% BSA with 0.09% Sodium Azide | |
| DnaJ (Hsp40) homolog, subfamily C, member 8, dnaJ homolog subfamily C member 8, HSPC331, SPF31Splicing protein spf31 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 22826 | |
| Store at 4C. Do not freeze. |
| Western Blot, Immunoprecipitation | |
| 0.2 mg/mL | |
| Western Blot 1:100-1:2000, Immunoprecipitation 1:10-1:500 | |
| NP_055095.2 | |
| DNAJC8 | |
| The immunogen recognized by this antibody maps to a region between residue 203 and 253 of human DnaJ (Hsp40) homolog, subfamily C, member 8 using the numbering given in entry NP_055095.2 (GeneID 22826). | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction