Learn More
Invitrogen™ DNAJC7 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA596207
Description
Immunogen sequence: VQFFVQALRM APDHEKACIA CRNAKALKAK KEDGNKAFKE GNYKLAYELY TEALGIDPNN IKTNAKLYCN RGTVNSKLRK LDDAIEDCTN AVKLDDTYIK AYLRRAQCYM DTEQYEEAVR DYEKVYQTEK TKEHKQLLKN AQLELKKSKR K; Positive Samples: A-431, 293T, K-562; Cellular Location: Cytoplasm, Nucleus, cytoskeleton.
DNAJC7 belongs to the evolutionarily conserved DNAJ/HSP40 family of proteins, which regulate molecular chaperone activity by stimulating ATPase activity. DNAJ proteins may have up to 3 distinct domains: a conserved 70-amino acid J domain, usually at the N terminus; a glycine/phenylalanine (G/F)-rich region; and a cysteine-rich domain containing 4 motifs resembling a zinc finger domain (Ohtsuka and Hata, 2000 ).
Specifications
| DNAJC7 | |
| Polyclonal | |
| Unconjugated | |
| DNAJC7 | |
| 2010003F24Rik; 2010004G07Rik; CCRP; Cytoplasmic CAR retention protein; cytoplasmic constitutive active/androstane receptor retention protein; DJ11; DJC7; DnaJ (Hsp40) homolog, subfamily C, member 7; DnaJ heat shock protein family (Hsp40) member C7; dnaJ homolog subfamily C member 7; DNAJC7; MDj11; mTpr2; tetratricopeptide repeat domain 2; tetratricopeptide repeat protein 2; TPR repeat protein 2; TPR2; TTC2 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 7266 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 1.51 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q99615 | |
| DNAJC7 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 230-380 of human DNAJC7 (NP_0033063). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.