missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
DNAJC30 Polyclonal specifically detects DNAJC30 in Human samples. It is validated for ELISA.
Specifications
Specifications
| Antigen | DNAJC30 |
| Applications | ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | DyLight 550 |
| Dilution | ELISA |
| Formulation | 50mM Sodium Borate |
| Gene Alias | DnaJ (Hsp40) homolog, subfamily C, member 30, dnaJ homolog subfamily C member 30, MGC12943, WBSCR18, Williams Beuren syndrome chromosome region 18, Williams-Beuren syndrome chromosomal region 18 protein |
| Host Species | Rabbit |
| Immunogen | Produced in rabbits immunized with purified, recombinant DNAJC30 (Q96LL9; Ser39-Gly124). |
| Purification Method | Affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?