Learn More
Invitrogen™ DNAJC28 Polyclonal Antibody

Chicken Polyclonal Antibody
Brand: Invitrogen™ PA19539
Description
PA1-9539 detects C21 in human samples. PA1-9539 has been successfully used in Western blot procedures. By Western blot, PA1-9539 detects a 40 kDa protein representing C21. PA1-9539 immunogen is a purified recombinant C21 corresponding to the chromosome 21 gene (Human).
Specifications
| DNAJC28 | |
| Polyclonal | |
| Unconjugated | |
| DNAJC28 | |
| BC020175; C21; C21orf55; C21orf78; DC42; DnaJ (Hsp40) homolog, subfamily C, member 28; DnaJ heat shock protein family (Hsp40) member C28; dnaJ homolog subfamily C member 28; DNAJC28; FLJ12894; IRA1; ORF28; TBLR2 | |
| Chicken | |
| IgY fraction | |
| RUO | |
| 54943 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide | |
| Q9NX36 | |
| DNAJC28 | |
| Purified recombinant C21 corresponding to the chromosome 21 gene (Human). | |
| 500 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgY |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.