missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DNAJC28 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA556332
This item is not returnable.
View return policy
Description
Immunogen sequence: RSHLIKATVI PNRVKMLPYF GIIRNRMMST HKSKKKIREY YRLLNVEEGC SADEVRESFH KLAKQYHPDS GSN Highest antigen sequence identity to the following orthologs: Mouse - 64%, Rat - 62%.
Specifications
| DNAJC28 | |
| Polyclonal | |
| Unconjugated | |
| DNAJC28 | |
| BC020175; C21; C21orf55; C21orf78; DC42; DnaJ (Hsp40) homolog, subfamily C, member 28; DnaJ heat shock protein family (Hsp40) member C28; dnaJ homolog subfamily C member 28; DNAJC28; FLJ12894; IRA1; ORF28; TBLR2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 54943 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9NX36 | |
| DNAJC28 | |
| Recombinant protein corresponding to Human DNAJC28. Recombinant protein control fragment (Product #RP-95902). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction