missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DNAJC25 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA569340
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 93%; Dog: 100%; Guinea Pig: 86%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 93%; Rat: 100%; Zebrafish: 77%.
This gene represents naturally-occurring mRNAs that are co-transcribed products of the neighboring DNAJC25 and GNG10 genes. These transcripts include the first exon of DNAJC25 and the last two exons of GNG10, resulting in a protein that combines the N-terminus of DNAJC25 and the C-terminus of GNG10. [provided by RefSeq, Dec 2008].
Specifications
| DNAJC25 | |
| Polyclonal | |
| Unconjugated | |
| DNAJC25 | |
| 2010109C08Rik; 2010203O07Rik; bA16L21.2.1; DnaJ (Hsp40) homolog, subfamily C , member 25; DnaJ (Hsp40) homolog, subfamily C, member 25; DnaJ heat shock protein family (Hsp40) member C25; dnaJ homolog subfamily C member 25; DNAJC25; DnaJ-like protein; dnaJ-like protein subfamily c member 25-like protein; ERJ7; GNG10; si:ch211-154e10.2 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 72429 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| A2ALW5 | |
| DNAJC25 | |
| synthetic peptide directed towards the middle region of human DNAJC25 (aa 173-222). | |
| 100 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction