missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DNAJC25 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA553929
This item is not returnable.
View return policy
Description
Immunogen sequence: NIKGKEYGEE ERLYIIRKSM KMSKSQFDSL EDHQKETFLK RELWIKENYE VYKQEQEEEL KKKLANDPRW KRYRRWMKNE GPGRLTFVDD Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 98%.
This gene represents naturally-occurring mRNAs that are co-transcribed products of the neighboring DNAJC25 and GNG10 genes. These transcripts include the first exon of DNAJC25 and the last two exons of GNG10, resulting in a protein that combines the N-terminus of DNAJC25 and the C-terminus of GNG10. [provided by RefSeq, Dec 2008].
Specifications
| DNAJC25 | |
| Polyclonal | |
| Unconjugated | |
| DNAJC25 | |
| 2010109C08Rik; 2010203O07Rik; bA16L21.2.1; DnaJ (Hsp40) homolog, subfamily C , member 25; DnaJ (Hsp40) homolog, subfamily C, member 25; DnaJ heat shock protein family (Hsp40) member C25; dnaJ homolog subfamily C member 25; DNAJC25; DnaJ-like protein; dnaJ-like protein subfamily c member 25-like protein; ERJ7; GNG10; si:ch211-154e10.2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 548645 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.5 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9H1X3 | |
| DNAJC25 | |
| Recombinant protein corresponding to Human DNAJC25. Recombinant protein control fragment (Product #RP-102372). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction