missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DNAJC25-GNG10 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£381.00
Specifications
| Antigen | DNAJC25-GNG10 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
DNAJC25-GNG10 Polyclonal specifically detects DNAJC25-GNG10 in Human samples. It is validated for Western Blot.Specifications
| DNAJC25-GNG10 | |
| Polyclonal | |
| Rabbit | |
| NP_004116 | |
| 552891 | |
| Synthetic peptide directed towards the N terminal of human LOC552891. Peptide sequence AGALVEGLYCGTRDCYEVLGVSRSAGKAEIARAYRQLARRYHPDRYRPQP. | |
| Primary |
| Western Blot | |
| Unconjugated | |
| RUO | |
| DNAJC25-GNG10 readthrough | |
| DNAJC25-GNG10 | |
| IgG | |
| 16 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title