missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
DNAJC2 Polyclonal antibody specifically detects DNAJC2 in Human samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | DNAJC2 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | DyLight 488 |
| Dilution | Western Blot |
| Formulation | 50mM Sodium Borate |
| Gene Alias | DnaJ (Hsp40) homolog, subfamily C, member 2, dnaJ homolog subfamily C member 2, MPHOSPH11M-phase phosphoprotein 11, MPP11Zuotin-related factor 1, ZRF1zuotin, ZUO1, zuotin related factor 1 |
| Host Species | Rabbit |
| Immunogen | The immunogen this antibody was made to, maps to a region between residue 571 and 621 of human DnaJ (Hsp40) Homolog, Subfamily C, Member 2 using the numbering given in entry NP_055192.1 (GeneID 27000). |
| Purification Method | Immunogen affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?