Learn More
DNAJC19 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 12096-1-AP-150UL
 View more versions of this product
Product Code. 16889603
  • £349.00 / 150µL
Description
This antibody is specific to DNAJC19 and had been tested by siRNA.
The protein encoded by this gene is thought to be part of a complex involved in the ATP-dependent transport of transit peptide-containing proteins from the inner cell membrane to the mitochondrial matrix. Defects in this gene are a cause of 3-methylglutaconic aciduria type 5 (MGA5), also known as dilated cardiomyopathy with ataxia (DCMA). Alternative splicing of this gene results in multiple transcript variants. Related pseudogenes have been identified on chromosomes 1, 2, 6, 10, 14 and 19.
Specifications
Specifications
DNAJC19
Polyclonal
Unconjugated
DNAJC19
DNAJC19, TIM14, TIMM14
Rabbit
Antigen Affinity Chromatography
RUO
131118, 502525, 67713
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot
0.21 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q96DA6, Q9CQV7
Dnajc19
DNAJC19 Fusion Protein Ag2739
150 μL
Primary
Human, Mouse, Rat