missing translation for 'onlineSavingsMsg'
Learn More

DNAJC19 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

£145.00 - £417.00

Specifications

Antigen DNAJC19
Dilution Western Blot 1:500 - 1:2000, ELISA, Immunocytochemistry/Immunofluorescence 1:50 - 1:200
Applications ELISA, Western Blot, Immunocytochemistry/Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30230951
View Documents
Novus Biologicals
NBP3-35489-20ul
20 μL
£145.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30229912
View Documents
Novus Biologicals
NBP3-35489-100ul
100 μL
£417.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

DNAJC19 Polyclonal antibody specifically detects DNAJC19 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot,Immunocytochemistry/Immunofluorescence
Specifications

Specifications

DNAJC19
ELISA, Western Blot, Immunocytochemistry/Immunofluorescence
Unconjugated
Rabbit
Vision
PBS (pH 7.3), 50% glycerol
131118
IgG
Affinity purified
Western Blot 1:500 - 1:2000, ELISA, Immunocytochemistry/Immunofluorescence 1:50 - 1:200
Polyclonal
Purified
RUO
Human, Mouse, Rat
DnaJ (Hsp40) homolog, subfamily C, member 19, DnaJ homolog subfamily C member 19, homolog of yeast TIM14, mitochondrial import inner membrane translocase subunit TIM14, Tim14, TIMM14TIM14Pam18
Recombinant fusion protein containing a sequence corresponding to amino acids 1-116 of human DNAJC19 (NP_660304.1).,, Sequence:, MASTVVAVGLTIAAAGFAGRYVLQAMKHMEPQVKQVFQSLPKSAFSGGYYRGGFEPKMTKREAALILGVSPTANKGKIRDAHRRIMLLNHPDKGGSPYIAAKINEAKDLLEGQAKK
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.