missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DNAJC17 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA528045
This item is not returnable.
View return policy
Description
Recommended positive controls: 293T, A431, H1299, HeLaS3, HepG2, Molt-4, Raji. Predicted reactivity: Mouse (92%), Rat (92%), Chicken (81%), Bovine (89%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
May negatively affect PAX8-induced thyroglobulin/TG transcription.
Specifications
| DNAJC17 | |
| Polyclonal | |
| Unconjugated | |
| DNAJC17 | |
| 1700025B16Rik; C87112; D9Bwg1371e; DnaJ (Hsp40) homolog, subfamily C, member 17; DnaJ heat shock protein family (Hsp40) member C17; dnaJ homolog subfamily C member 17; Dnajc17; RGD1308015 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 55192 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 0.7 mg/mL | |
| 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
| Q9NVM6 | |
| DNAJC17 | |
| Recombinant protein encompassing a sequence within the center region of human DNAJC17. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction