missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DNAJC17 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5101267
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total DNAJC17.
May negatively affect PAX8-induced thyroglobulin/TG transcription.
Specifications
| DNAJC17 | |
| Polyclonal | |
| Unconjugated | |
| DNAJC17 | |
| 1700025B16Rik; C87112; D9Bwg1371e; DnaJ (Hsp40) homolog, subfamily C, member 17; DnaJ heat shock protein family (Hsp40) member C17; dnaJ homolog subfamily C member 17; Dnajc17; RGD1308015 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 55192, 69408 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q91WT4, Q9NVM6 | |
| DNAJC17 | |
| A synthesized peptide derived from human DNAJC17(Accession Q9NVM6), corresponding to amino acid residues E19-R69. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction