missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DNAJC15 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA550418
This item is not returnable.
View return policy
Description
The antibody detects endogenous levels of total DNAJC15 protein.
DNAJC15 is a negative regulator of the mitochondrial respiratory chain. DNAJC15 prevents mitochondrial hyperpolarization state and restricts mitochondrial generation of ATP. DNAJC15 also acts as an import component of the TIM23 translocase complex, and stimulates the ATPase activity of HSPA9. Diseases associated with DNAJC15 include Cicatricial Entropion.
Specifications
| DNAJC15 | |
| Polyclonal | |
| Unconjugated | |
| DNAJC15 | |
| 1110003P16Rik; cell growth-inhibiting gene 22 protein; DnaJ (Hsp40) homolog, subfamily C, member 15; DnaJ (Hsp40) homolog, subfamily D, member 1; DNAJ domain-containing; DnaJ heat shock protein family (Hsp40) member C15; dnaJ homolog subfamily C member 15; DNAJC15; Dnajd1; GIG22; HSD18; MCJ; methylation-controlled J protein | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 29103 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1.5 mg/mL | |
| PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| Q9Y5T4 | |
| DNAJC15 | |
| Fusion protein corresponding to a region derived from internal residues of human DnaJ (Hsp40) homolog, subfamily C, member 15. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction