missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DNAJC14 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 2 publications
£222.00 - £443.00
Specifications
| Antigen | DNAJC14 |
|---|---|
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18457111
|
Novus Biologicals
NBP1-90488-25ul |
25 μL |
£222.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18262498
|
Novus Biologicals
NBP1-90488 |
0.1 mL |
£443.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
DNAJC14 Polyclonal specifically detects DNAJC14 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| DNAJC14 | |
| Polyclonal | |
| Rabbit | |
| Human, Mouse, Rat | |
| DnaJ (Hsp40) homolog, subfamily C, member 14, dnaJ homolog subfamily C member 14, dnaJ protein homolog 3, dopamine receptor interacting protein, dopamine receptor-interacting protein of 78 kDa, hDj-3, HDJ3, human DnaJ protein 3, LIP6, LYST-interacting protein LIP6 | |
| DNAJC14 | |
| IgG | |
| Affinity Purified | |
| Specificity of human DNAJC14 antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunofluorescence | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 85406 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:WGWLELPWVKQNINRQGNAPVASGRYCQPEEEVARLLTMAGVPEDELNPFHVLGVEATASDVELKKAYRQLAVMVHPDKNHHPRAEEAFKVLRAAWDIVSNAEKRKEYEMKRMAENELSRSVNEFLSKLQDDLKEAMNTMMCSRCQG | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title