missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DNAJC13 Recombinant Rabbit Monoclonal Antibody (11H13L5)
DNAJC13 Recombinant Rabbit Monoclonal Antibody (11H13L5)
Brand: Invitrogen 702773
This item is not returnable.
View return policy
Description
DNAJC13 Recombinant Monoclonal Antibody for Western Blot, ICC/IF
Specifications
| DNAJC13 | |
| Recombinant Monoclonal | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| O75165 | |
| DNAJC13 | |
| Protein corresponding to human DNAJC13 (aa1301-aa1354) GST Fusion. | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
| Western Blot, Immunocytochemistry | |
| 11H13L5 | |
| Unconjugated | |
| DNAJC13 | |
| D030002L11Rik; DnaJ (Hsp40) homolog, subfamily C, member 13; DnaJ domain-containing protein RME-8; DnaJ heat shock protein family (Hsp40) member C13; dnaJ homolog subfamily C member 13; DNAJC13; Gm1124; j-domain containing protein, required for endocytosis; KIAA0678; mKIAA0678; PARK21; receptor mediated endocytosis RME-8; required for receptor-mediated endocytosis 8; Rme8; RME-8 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 23317, 235567 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction