missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DNAJC13 Antibody, mFluor Violet 610 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-81919MFV610
This item is not returnable.
View return policy
Description
DNAJC13 Polyclonal antibody specifically detects DNAJC13 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| DNAJC13 | |
| Polyclonal | |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| DnaJ (Hsp40) homolog, subfamily C, member 13, DnaJ domain-containing protein RME-8, dnaJ homolog subfamily C member 13, required for receptor-mediated endocytosis 8 | |
| DNAJC13 antibody was raised against an 18 amino acid peptide near the carboxy terminus of human DNAJC13. The immunogen is located within amino acids 1940 - 1990 of DNAJC13. Amino Acid Squence: EHFKNQQDNPEANWKLPE | |
| 0.1 mL | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| 23317 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction