missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
DNAJC13 Polyclonal antibody specifically detects DNAJC13 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
Specifications
| Immunogen | DNAJC13 antibody was raised against an 18 amino acid peptide near the carboxy terminus of human DNAJC13. The immunogen is located within amino acids 1940 - 1990 of DNAJC13. Amino Acid Squence: EHFKNQQDNPEANWKLPE |
| Gene ID (Entrez) | 23317 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?