missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ DNAJC12 Recombinant Rabbit Monoclonal Antibody (K01_2M15)

Product Code. 30829532
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30829532 50 μL
30830120 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30829532 Supplier Invitrogen™ Supplier No. MA565506
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

DNAJC12 Recombinant Monoclonal Antibody for Western Blot

This gene encodes a member of a subclass of the HSP40/DnaJ protein family. Members of this family of proteins are associated with complex assembly, protein folding, and export. Two transcript variants encoding distinct isoforms have been identified for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DNAJC12
Applications Western Blot
Classification Recombinant Monoclonal
Clone K01_2M15
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 50mM tris glycine with 0.05% BSA, 40% glycerol, 150mM NaCl and 0.01% sodium azide; pH 7.4
Gene DNAJC12
Gene Accession No. Q9UKB3
Gene Alias DnaJ (Hsp40) homolog, subfamily C, member 12; DnaJ heat shock protein family (Hsp40) member C12; dnaJ homolog subfamily C member 12; DnaJ subfamily C member 12-like protein; DNAJC12; DnaJ-like protein subfamily C member 12-like protein; J domain containing protein 1; J domain containing protein 1 (JDP1); J domain protein 1; j domain-containing protein 1; JDP1; mJDP1
Gene Symbols DNAJC12
Host Species Rabbit
Immunogen A synthetic peptide of human DNAJC12.
Purification Method Protein A/G
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56521
Target Species Human
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.