missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DNAJC12 Antibody (S04-3E8), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-19858-100ul
This item is not returnable.
View return policy
Description
DNAJC12 Monoclonal antibody specifically detects DNAJC12 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| DNAJC12 | |
| Monoclonal | |
| 0.3 mg/mL | |
| 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| S04-3E8 | |
| Unconjugated | |
| DnaJ (Hsp40) homolog, subfamily C, member 12, dnaJ homolog subfamily C member 12, J domain containing protein 1 (JDP1), J domain protein 1, JDP1J domain-containing protein 1 | |
| A synthetic peptide of human DNAJC12 (Uniprot # Q9UKB3) | |
| 100 μg | |
| Signal Transduction | |
| 56521 | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction