missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DNAJC11 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5100479
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total DNAJC11.
Belongs to the DNAJC11 family. Three named isoforms produced by alternative splicing.
Specifications
| DNAJC11 | |
| Polyclonal | |
| Unconjugated | |
| Dnajc11 | |
| dJ126A5.1; DnaJ (Hsp40) homolog, subfamily C, member 11; DnaJ heat shock protein family (Hsp40) member C11; dnaJ homolog subfamily C member 11; DNAJC11; E030019A03Rik; novel DnaJ domain-containing protein | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 230935, 362666, 55735 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q5U458, Q9NVH1 | |
| Dnajc11 | |
| A synthesized peptide derived from human DNAJC11(Accession Q9NVH1), corresponding to amino acid residues K484-V534. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat, Monkey | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction