missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DNAJC10 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA543346
This item is not returnable.
View return policy
Description
Peptide sequence: DFYSLLGVSK TASSREIRQA FKKLALKLHP DKNPNNPNAH GDFLKINRAY Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 93%; Horse: 93%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 83%.
This endoplasmic reticulum co-chaperone may play a role in protein folding and translocation across the endoplasmic reticulum membrane. DNAJC10 may act as a co-chaperone for HSPA5.
Specifications
| DNAJC10 | |
| Polyclonal | |
| Unconjugated | |
| Dnajc10 | |
| 1200006L06Rik; D2Ertd706e; DnaJ (Hsp40) homolog, subfamily C, member 10; DnaJ heat shock protein family (Hsp40) member C10; dnaJ homolog subfamily C member 10; DNAJC10; DnaJ-like protein subfamily C member 10-like protein; Endoplasmic reticulum DNA J domain-containing protein 5; endoplasmic reticulum DnaJ-PDI fusion protein 1; ERdj5; ER-resident protein ERdj5; J domain-containing PDI-like protein; j domain-containing protein disulfide isomerase-like protein; J-domain-containing protein disulfide isomerase-like protein; Jpdi; Macrothioredoxin; MTHr; PDIA19; protein disulfide isomerase family A, member 19; UNQ495/PRO1012; zgc:162218 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 54431 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q8IXB1 | |
| Dnajc10 | |
| Synthetic peptide directed towards the N-terminal of human DNAJC10 (aa 35-84). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction