Learn More
Invitrogen™ DNAJB9 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA596702
Description
Immunogen sequence: SKSYYDILGV PKSASERQIK KAFHKLAMKY HPDKNKSPDA EAKFREIAEA YETLSDANRR KEYDTLGHSA FTSGKGQRGS GSSFEQSFNF NFDDLFKDFG FFGQNQNTGS KKRFENHFQT RQDGGSSRQR HHFQEFSFGG GLFDDMFEDM EKMFSFSGFD STNQHTVQTE NRFHGSSKHC RTVTQRRGNM VTTYTDCSGQ; Positive Samples: Mouse liver, Rat liver; Cellular Location: Endoplasmic reticulum lumen.
This gene is a member of the J protein family. J proteins function in many cellular processes by regulating the ATPase activity of 70 kDa heat shock proteins. This gene is a member of the type 2 subgroup of DnaJ proteins. The encoded protein is localized to the endoplasmic reticulum. This protein is induced by endoplasmic reticulum stress and plays a role in protecting stressed cells from apoptosis.
Specifications
| DNAJB9 | |
| Polyclonal | |
| Unconjugated | |
| DNAJB9 | |
| AA408011; AA673251; AA673481; AW556981; DKFZP564F1862; DNA j protein b9; DnaJ (Hsp40) homolog, subfamily B, member 9; DnaJ heat shock protein family (Hsp40) member B9; dnaJ homolog subfamily B member 9; dnaJ homolog, subfamily b, member 9; DNAJB9; Endoplasmic reticulum DNA J domain-containing protein 4; endoplasmic reticulum DnaJ homolog 4; ERdj4; ER-resident protein ERdj4; Mdg1; MDG-1; mDj7; microvascular endothelial differentiation gene 1; microvascular endothelial differentiation gene 1 protein; MST049; MSTP049; UNQ743/PRO1471 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 24908, 27362 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.87 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P97554, Q9QYI6 | |
| DNAJB9 | |
| Recombinant protein (or fragment) | |
| 100 μL | |
| Primary | |
| Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.