missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DNAJB9 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA559621
This item is not returnable.
View return policy
Description
Immunogen sequence: TLSDANRRKE YDTLGHSAFT SGKGQRGSGS SFEQSFNFNF DDLFKDFGFF GQNQNTGSKK RFENHFQTRQ DG Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 86%.
This gene is a member of the J protein family. J proteins function in many cellular processes by regulating the ATPase activity of 70 kDa heat shock proteins. This gene is a member of the type 2 subgroup of DnaJ proteins. The encoded protein is localized to the endoplasmic reticulum. This protein is induced by endoplasmic reticulum stress and plays a role in protecting stressed cells from apoptosis.
Specifications
| DNAJB9 | |
| Polyclonal | |
| Unconjugated | |
| DNAJB9 | |
| AA408011; AA673251; AA673481; AW556981; DKFZP564F1862; DNA j protein b9; DnaJ (Hsp40) homolog, subfamily B, member 9; DnaJ heat shock protein family (Hsp40) member B9; dnaJ homolog subfamily B member 9; dnaJ homolog, subfamily b, member 9; DNAJB9; Endoplasmic reticulum DNA J domain-containing protein 4; endoplasmic reticulum DnaJ homolog 4; ERdj4; ER-resident protein ERdj4; Mdg1; MDG-1; mDj7; microvascular endothelial differentiation gene 1; microvascular endothelial differentiation gene 1 protein; MST049; MSTP049; UNQ743/PRO1471 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 4189 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunohistochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9UBS3 | |
| DNAJB9 | |
| Recombinant protein corresponding to Human DNAJB9. Recombinant protein control fragment (Product #RP-98421). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction