missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DNAJB8 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA524140
This item is not returnable.
View return policy
Description
DNAJB8 Polyclonal Antibody for Western Blot
DNAJB8 belongs to the evolutionarily conserved DNAJ/HSP40 family of proteins, which regulate molecular chaperone activity by stimulating ATPase activity. DNAJ proteins may have up to 3 distinct domains: a conserved 70-amino acid J domain, usually at the N terminus; a glycine/phenylalanine (G/F)-rich region; and a cysteine-rich domain containing 4 motifs resembling a zinc finger domain.
Specifications
| DNAJB8 | |
| Polyclonal | |
| Unconjugated | |
| DNAJB8 | |
| 1700016F14Rik; 1700031F10Rik; AI326313; CT156; DJ6; DnaJ (Hsp40) homolog, subfamily B, member 8; DnaJ heat shock protein family (Hsp40) member B8; DnaJ homolog 6; dnaJ homolog subfamily B member 8; DNAJB8; mDj6; RGD1561981; testicular tissue protein Li 56 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 165721, 56691 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q8NHS0, Q9QYI7 | |
| DNAJB8 | |
| KLH conjugated synthetic peptide between 41-69 amino acids from the N-terminal region of human DNAJB8. | |
| 400 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction