missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DNAJB6 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA563592
This item is not returnable.
View return policy
Description
Immunogen sequence: VADDDALAEE RMRRGQNALP AQPAGLRPPK PPRPASLLRH APHCLSEEEG EQDRPRAPGP WDPLASAAGL K Highest antigen sequence identity to the following orthologs: Mouse - 33%, Rat - 37%.
This gene encodes a member of the DNAJ protein family. DNAJ family members are characterized by a highly conserved amino acid stretch called the 'J-domain' and function as one of the two major classes of molecular chaperones involved in a wide range of cellular events, such as protein folding and oligomeric protein complex assembly. This family member may also play a role in polyglutamine aggregation in specific neurons. Alternative splicing of this gene results in multiple transcript variants; however, not all variants have been fully described.
Specifications
| DNAJB6 | |
| Polyclonal | |
| Unconjugated | |
| DNAJB6 | |
| DJ4; DnaJ; DnaJ (Hsp40) homolog, subfamily B, member 6; DnaJ heat shock protein family (Hsp40) member B6; dnaJ homolog subfamily B member 6; DNAJB6; dnajb6 {ECO:0000312; DnaJ-like 2 protein; EMBL:AAH78908.1, ECO:0000312; Heat shock protein J2; HHDJ1; Hsj2; HSJ-2; Hsp40 homolog; LGMD1D; LGMD1E; mammalian relative of DnaJ; mDj4; MRJ; MSJ1; MSJ-1; RGD:1308207} | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 10049 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O75190 | |
| DNAJB6 | |
| Recombinant protein corresponding to Human DNAJB6. Recombinant protein control fragment (Product #RP-103894). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction