missing translation for 'onlineSavingsMsg'
Learn More

DNAJB5 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

£135.00 - £349.00

Specifications

Antigen DNAJB5
Concentration 0.16 mg/mL
Applications Immunocytochemistry, Western Blot, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16886074
View Documents
Proteintech
16453-1-AP-20UL
20 μL
£135.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16876074
View Documents
Proteintech
16453-1-AP-150UL
150 μL
£349.00
150µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

DNAJB5 belongs to the evolutionarily conserved DNAJ/HSP40 protein family. For background information on the DNAJ family.
Specifications

Specifications

DNAJB5
Immunocytochemistry, Western Blot, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation
Unconjugated
Rabbit
Human, Mouse, Rat
O75953, O89114
25822, 313811, 56323
DNAJB5 Fusion Protein Ag9341
Primary
-20°C
DNAJB5
0.16 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
DNAJB5, Heat shock protein cognate 40, Heat shock protein Hsp40 2, Heat shock protein Hsp40 3, Hsc40, KIAA1045
Dnajb5
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.