missing translation for 'onlineSavingsMsg'
Learn More

DNAJB5 Antibody (1C3), Novus Biologicals™

Product Code. 18395699 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18395699 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18395699 Supplier Novus Biologicals Supplier No. H00025822M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

DNAJB5 Monoclonal antibody specifically detects DNAJB5 in Human samples. It is validated for Western Blot, ELISA, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen DNAJB5
Applications Western Blot, ELISA, Sandwich ELISA
Classification Monoclonal
Clone 1C3
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA, Sandwich ELISA
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH12115
Gene Alias DnaJ (Hsp40) homolog, subfamily B, member 5, heat shock cognate 40, Heat shock protein cognate 40, Heat shock protein Hsp40-2, Heat shock protein Hsp40-3, HSC40, Hsc40dnaJ homolog subfamily B member 5, KIAA1045
Host Species Mouse
Immunogen DNAJB5 (AAH12115, 1 a.a. ~ 386 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MGKDYYKILGIPSGANEDEIKKAYRKMALKYHPDKNKEPNAEEKFKEIAEAYDVLSDPKKRGLYDQYGEEGLKTGGGTSGGSSGSFHYTFHGDPHATFASFFGGSNPFDIFFASSRSTRPFSGFDPDDMDVDEDEDPFGAFGRFGFNGLSRGPRRAPEPLYPRRKVQDPPVVHELRVSLEEIYHGSTKRMKITRRRLNPDGRTVRTEDKILHIVIKRGWKEGTKITFPKEGDATPDNIPADIVFVLKDKPHAHFRRDGTNVLYSALISLKEALCGCTVNIPTIDGRVIPLPCNDVIKPGTVKRLRGEGLPFPKVPTQRGDLIVEFKVRFPDRLTPQTRQILKQQYPQHSLHSMCTQLDVHWTLATFSKMQKKATGFQENVPVPDPF
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25822
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.