missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ DNAJB3 Polyclonal Antibody
GREENER_CHOICE

Product Code. 16636584
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16636584 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16636584 Supplier Invitrogen™ Supplier No. PA5100096

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total DNAJB3.

May operate as a co-chaperone of the male germ cell- and haploid stage-specific Hsp70 proteins. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen DNAJB3
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene DNAJB3
Gene Accession No. O35723, Q8WWF6
Gene Alias DnaJ (Hsp40) homolog, subfamily B, member 3; DnaJ heat shock protein family (Hsp40) member B3; dnaJ homolog subfamily B member 3; DnaJ Hsp40 homolog; DnaJ protein homolog 3; DNAJB3; HCG3; heat shock protein J3; heat shock protein, DNAJ-like 3; Hsj3; HSJ-3; Msj1; MSJ-1
Gene Symbols DNAJB3
Host Species Rabbit
Immunogen A synthesized peptide derived from human DNAJB3(Accession Q8WWF6), corresponding to amino acid residues D85-Q135.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 15504, 414061, 680216
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.