missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DNAJB2, Mouse anti-Human, Clone: 2C9, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00003300-M03.100ug
This item is not returnable.
View return policy
Description
Sequence: LELSRREQQPSVTSRSGGTQVQQTPASCPLDSDLSEDEDLQLAMAYSLSEMEAAGKKPAGGREAQHRRQGRPKAQHQDPGLGGTQEGARGEATKRSPSPEEKASRCLILSpecifications
| DNAJB2 | |
| Monoclonal | |
| Unconjugated | |
| DnaJ (Hsp40) homolog, subfamily B, member 2 | |
| HSJ1/HSPF3 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 2C9 | |
| In 1x PBS, pH 7.4 | |
| NM_006736 | |
| DNAJB2 | |
| DNAJB2 (NP_006727, 216 a.a. ∼ 324 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 3300 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction