missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DNAJB14 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97528-50ul
This item is not returnable.
View return policy
Description
DNAJB14 Polyclonal antibody specifically detects DNAJB14 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| DNAJB14 | |
| Polyclonal | |
| Immunohistochemistry-Paraffin 1:100-1:400 | |
| DnaJ (Hsp40) homolog, subfamily B, member 14, dnaJ homolog subfamily B member 14, EGNR9427, FLJ14281, MGC22187, PRO34683 | |
| Produced in rabbits immunized with E. coli-derived Human DNAJB14 fragment. | |
| 50 μg | |
| Signal Transduction | |
| 79982 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.0) | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction