missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DNAJB11 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA578564
This item is not returnable.
View return policy
Description
Predicted Reactivity: Human (100%), Zebrafish (83%), Xenopus laevis (87%), Pig (99%), Bovine (97%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
DNAJB11 belongs to the evolutionarily conserved DNAJ/HSP40 family of proteins, which regulate molecular chaperone activity bystimulating ATPase activity. DNAJ proteins may have up to 3 distinct domains: a conserved 70-amino acid J domain, usually at the N terminus a glycine/phenylalanine (G/F)-rich region and a C-terminal cysteine-rich region.
Specifications
| DNAJB11 | |
| Polyclonal | |
| Unconjugated | |
| DNAJB11 | |
| 1810031F23Rik; ABBP2; ABBP-2; AL024055; APOBEC1-binding protein 2; cb954; Dj9; Dj-9; DnaJ (Hsp40) homolog, subfamily B, member 11; DnaJ heat shock protein family (Hsp40) member B11; dnaJ homolog subfamily B member 11; DnaJ protein 9; DnaJ protein homolog 9; DNAJB11; dnajb11 protein; DnaJ-like protein subfamily B member 11-like protein; EDJ; Endoplasmic reticulum DNA J domain-containing protein 3; ER-associated DNAJ; ER-associated DNAJ protein 3; ER-associated Hsp40 co-chaperone; ERdj3; ERJ3; ERj3p; ER-resident protein ERdj3; HDJ9; hDj-9; HEDJ; Human DnaJ protein 9; Liver regeneration-related protein LRRGT00084; LRRGT00084; PRO1080; PSEC0121; PWP1-interacting protein 4; UNQ537; UNQ537/PRO1080 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 360734, 51726, 67838 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.89 mg/mL | |
| PBS with 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q6TUG0, Q99KV1, Q9UBS4 | |
| DNAJB11 | |
| Recombinant protein encompassing a sequence within the center region of human DNAJB11. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction