missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DNAJB11 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92708-0.1ml
This item is not returnable.
View return policy
Description
DNAJB11 Polyclonal antibody specifically detects DNAJB11 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| DNAJB11 | |
| Polyclonal | |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunohistochemistry-Paraffin | |
| ABBP-2, APOBEC1-binding protein 2, DnaJ (Hsp40) homolog, subfamily B, member 11, dnaJ homolog subfamily B member 11, DnaJ protein homolog 9, EDJABBP2, ER-associated DNAJ, ER-associated dnaJ protein 3, ER-associated Hsp40 co-chaperone, ERdj3DJ9, ERJ3, ERj3p, hDj9, hDj-9, HEDJERj3, Human DnaJ protein 9, PRO1080, PWP1-interacting protein 4, UNQ537 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 89-358 of human DNAJB11 (NP_057390.1). YGEEGLKDGHQSSHGDIFSHFFGDFGFMFGGTPRQQDRNIPRGSDIIVDLEVTLEEVYAGNFVEVVRNKPVARQAPGKRKCNCRQEMRTTQLGPGRFQMTQEVVCDECPNVKLVNEERTLEVEIEPGVRDGMEYPFIGEGEPHVDGEPGDLRFRIKVVKHPIFERRGDDLYTNVTISLVESLVGFEMDITHLDGHKVHISRDKITRPGAKLWKKGEGLPNFDNNNIKGSLIITFDVDFPKEQLTEEAREGIKQLLKQGSVQKVYNGLQGY | |
| 0.1 mL | |
| Signal Transduction | |
| 51726 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction