missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DNAJB1 Rabbit Recombinant Antibody, Proteintech
Rabbit Recombinant Antibody
Brand: Proteintech 82676-1-RR-100UL
This item is not returnable.
View return policy
Description
The DNAJB1 antibody from Proteintech is a rabbit recombinant antibody generated with recombinant protein of human DNAJB1. This antibody recognizes human, mouse, rat antigen. The DNAJB1 antibody has been validated for the following applications: WB, IHC, IF/ICC, IP, ELISA analysis.Specifications
| DNAJB1 | |
| Recombinant Monoclonal | |
| 1000 μg/mL | |
| HSPF1; Hdj1; Hsp40; Sis1 | |
| Recombinant Protein | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| 37-40 kDa | |
| Liquid |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, ELISA | |
| 4K3 | |
| Unconjugated | |
| Rabbit | |
| Protein A purification | |
| RUO | |
| 3337 | |
| Store at -20°C. Stable for one year after shipment. | |
| Recombinant Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction