missing translation for 'onlineSavingsMsg'
Learn More

DNAJA4 Antibody, Alexa Fluor™ 750, Novus Biologicals™

Product Code. 18411508 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18411508 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18411508 Supplier Novus Biologicals Supplier No. NBP281983AF750

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

DNAJA4 Polyclonal antibody specifically detects DNAJA4 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen DNAJA4
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate Alexa Fluor 750
Formulation 50mM Sodium Borate
Gene Alias DnaJ (Hsp40) homolog, subfamily A, member 4, dnaJ homolog subfamily A member 4, MST104, MSTP104, PRO1472
Host Species Rabbit
Immunogen DNAJA4 antibody was raised against a 15 amino acid peptide near the center of human DNAJA4. The immunogen is located within amino acids 210 - 260 of DNAJA4. Amino Acid Squence: HGEGDQEPELEPGDV
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55466
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.