missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
DNAJA2 Monoclonal antibody specifically detects DNAJA2 in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Flow Cytometry
Specifications
Specifications
| Antigen | DNAJA2 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | OTI3A10 |
| Conjugate | mFluor Violet 500 SE |
| Dilution | Flow Cytometry |
| Formulation | 50mM Sodium Borate |
| Gene Alias | cell cycle progression 3 protein, Cell cycle progression restoration gene 3 protein, CPR3PRO3015, Dj3, DJA2, DNAJ, DnaJ (Hsp40) homolog, subfamily A, member 2, dnaJ homolog subfamily A member 2, DNJ3, HIRA interacting protein 4, HIRA-interacting protein 4, HIRIP4RDJ2, Renal carcinoma antigen NY-REN-14 |
| Host Species | Mouse |
| Immunogen | Full length human recombinant protein of human DNAJA2 (NP_005871) produced in HEK293T cell. |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?