missing translation for 'onlineSavingsMsg'
Learn More

DNAI1 Rabbit anti-Mouse, Rat, Clone: 4Y6Q1, Novus Biologicals™

Product Code. 18317524 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18317524 20 μg 20µL
18392105 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18317524 Supplier Novus Biologicals Supplier No. NBP31623520UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

DNAI1 Monoclonal antibody specifically detects DNAI1 in Mouse, Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen DNAI1
Applications Western Blot
Classification Monoclonal
Clone 4Y6Q1
Conjugate Unconjugated
Dilution Western Blot 1:500 - 1:2000
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias Axonemal dynein intermediate chain 1, CILD1MGC26204, dynein intermediate chain DNAI1, dynein, axonemal, intermediate chain 1, dynein, axonemal, intermediate polypeptide 1, ICS, ICS1, immotile cilia syndrome 1, PCDdynein intermediate chain 1, axonemal
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-200 of human DNAI1 (Q9UI46). MIPASAKAPHKQPHKQSISIGRGTRKRDEDSGTEVGEGTDEWAQSKATVRPPDQLELTDAELKEEFTRILTANNPHAPQNIVRYSFKEGTYKPIGFVNQLAVHYTQVGNLIPKDSDEGRRQHYRDELVAGSQESVKVISETGNLEEDEEPKELETEPGSQTDVPAAGAAEKVTEEELMTPKQPKERKLTNQFNFSERASQ
Purification Method Affinity purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Cell Biology, Cytoskeleton Markers, Neurodegeneration, Neuroscience, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 27019
Target Species Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.