Learn More
Invitrogen™ DNAH9 Monoclonal Antibody (440.4)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA126770
Description
Recommended positive controls: chicken brain and kinesin enriched rat brain extract. MA1-26770 will not react with hamster CHO cell chromosomal dynein or dynein from Chlamydomonas flagella. By Western blot, MA1-26770 detects an ~440kD protein representing Dynein heavy chain. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes the heavy chain subunit of axonemal dynein, a large multi-subunit molecular motor. Axonemal dynein attaches to microtubules and hydrolyzes ATP to mediate the movement of cilia and flagella. The gene expresses at least two transcript variants; additional variants have been described, but their full length nature has not been determined.
Specifications
| DNAH9 | |
| Monoclonal | |
| Conc. Not Determined | |
| Ascites with 15mM sodium azide | |
| Q9NYC9 | |
| DNAH9 | |
| Cytoplasmic dynein from chicken embryo brain. | |
| RUO | |
| 1770, 417314 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Dot Blot, ELISA, Western Blot, Immunocytochemistry | |
| 440.4 | |
| Unconjugated | |
| DNAH9 | |
| 9030022M04; A230091C01; axonemal beta dynein heavy chain 9; Bv2; C230051G16; ciliary dynein heavy chain 9; D11Ertd686e; DNAH17L; Dnah9; DNAH9 variant protein; Dnahc9; DNAL1; DNEL1; DYH9; dynein axonemal heavy chain 9; dynein heavy chain; dynein heavy chain 9, axonemal; dynein heavy chain Bv2; dynein, axonemal, heavy chain 9; dynein, axonemal, heavy polypeptide 9; HL20; HL-20; KIAA0357; mKIAA0357 | |
| Mouse | |
| 100 μL | |
| Primary | |
| Chicken, Human | |
| Antibody | |
| IgG2a |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.