missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DNAH8 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5148367
This item is not returnable.
View return policy
Description
DNAH8 Polyclonal Antibody for IHC (P)
Specifications
| DNAH8 | |
| Polyclonal | |
| Unconjugated | |
| DNAH8 | |
| ATPase; Axonemal beta dynein heavy chain 8; ciliary dynein heavy chain 8; DNAH8; dynein axonemal heavy chain 8; dynein heavy chain 8, axonemal; dynein, axonemal, heavy chain 8; dynein, axonemal, heavy polypeptide 8; hdhc9 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 1769 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 100 μg/mL | |
| PBS with 0.03% ProClin 300; pH 7 | |
| Q96JB1 | |
| DNAH8 | |
| E. coli-derived Human DNAH8 fragment. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction