missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ DNAH5 Split FISH Probe
Labeled FISH probes for identification of gene split using Fluoresecent In Situ Hybridization Technique.
Brand: Abnova™ FS0062.200uL
Additional Details : Weight : 0.02000kg
Specifications
Store at 4°C in the dark | |
Fluorescent In Situ Hybridization (Cell) | |
CILD3,DNAHC5,FLJ46759,HL1,KIAA1603,KTGNR,PCD | |
1767 | |
200 μL |
Tube | |
dynein, axonemal, heavy chain 5 | |
DNAH5 | |
DNA/RNA | |
Liquid |